Learn More
ACROBiosystems Human CNR1 Full Length Protein (VLP)
Catalog No. 502331869
Encompass_Preferred
Human CNR1 Full Length Protein-VLP (CN1-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 472 (Accession # P21554-1 (T210A)).
Catalog No. 50-233-1869 Supplier ACROBiosystems Supplier No. CN1H52P620UG
Human CNR1 Full Length Protein-VLP (CN1-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 472 (Accession # P21554-1 (T210A)).