Learn More
Sino Biological Recombinant Human CNR1 Protein (VLP, Flag-Strep & His tag)
Catalog No. 502993062
Encompass_Preferred
A DNA sequence encoding the Human CNR1 (P21554-1) (Met1-Ser414) was expressed, with a Flag tag followed by a Strep Tag at the N-terminus, a polyhistidine tag at the C-terminus. This protein was prepared using our VLP platform.As an innovative technology platform, VLPs are generated through the self-assembly of viral proteins in HEK293 cells. This process efficiently integrates membrane proteins onto the VLP surface, preserving their native conformation and yielding soluble, high-concentration proteins that are ideally suited for immunization and antibody screening.
Catalog No. 50-299-3062 Supplier Sino Biological Supplier No. 17407H91H500
A DNA sequence encoding the Human CNR1 (P21554-1) (Met1-Ser414) was expressed, with a Flag tag followed by a Strep Tag at the N-terminus, a polyhistidine tag at the C-terminus. This protein was prepared using our VLP platform.As an innovative technology platform, VLPs are generated through the self-assembly of viral proteins in HEK293 cells. This process efficiently integrates membrane proteins onto the VLP surface, preserving their native conformation and yielding soluble, high-concentration proteins that are ideally suited for immunization and antibody screening.