Learn More
Sino Biological Anti-CNTF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11841RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CNTF / Ciliary Neurotrophic Factor (rh CNTF; Catalog#11841-H07E; NP_000605.1; Ala2-Met200). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9119