Learn More
FUJIFILM Biosciences Human CNTF, Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Catalog No. 50990757
Encompass_Preferred
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.
Catalog No. 50-990-757 Supplier FUJIFILM Biosciences Supplier No. 100595UG
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.