Learn More
Gibco™ Human CNTF Recombinant Protein, PeproTech®
Description
Specifications
Specifications
| Accession Number | P26441 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 1270 |
| Molecular Weight (g/mol) | 22.8 kDa |
| Name | CNTF |
| Quantity | 250 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.