Learn More
Sino Biological Anti-Contactin-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10178T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CNTN4 (rh CNTN4; Catalog#10178-H08B; Q8IWV2-1; Met1-Ser1000). CNTN4 specific IgG was purified by Human CNTN4 affinity chromatography.
Catalog No. 50-240-9562