Learn More
Sino Biological Anti-CNTN5 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10513MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CNTN5 / Contactin-5 (rhCNTN5; Catalog#10513-H08H; NP_055176.1; Met 1-Gln 1059). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8653