Learn More
Biomatik Corporation Human Coactosin Like Protein 1 (COTL1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12073
Encompass_Preferred
Human Coactosin Like Protein 1 (COTL1) Mini sample ELISA Kit for detecting COTL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14019. Gene ID: 23406. Target Synonyms: CLP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8041