Learn More
ACROBiosystems Human Coagulation Factor III / Tissue Factor Protein, Fc Tag
Catalog No. 502054712
Encompass_Preferred
Recombinant Protein;100UG;Human Tissue Factor, Fc Tag (TF3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 251 (Accession # P13726-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.7 kDa.
Catalog No. 50-205-4712 Supplier ACROBiosystems Supplier No. TF3H5253100UG
Recombinant Protein;100UG;Human Tissue Factor, Fc Tag (TF3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 251 (Accession # P13726-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.7 kDa.