Learn More
ACROBiosystems ACROBiosystems Human Coagulation factor VII / F7 Protein, His Tag (active enzyme, MALS verified)
Catalog No. 502106948
Encompass_Preferred
Recombinant Protein;20UG;Human Coagulation factor VII, His Tag (TF7-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 61 - Pro 466 (Accession # P08709-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6948 Supplier ACROBiosystems Supplier No. TF7H52H520UG
Recombinant Protein;20UG;Human Coagulation factor VII, His Tag (TF7-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 61 - Pro 466 (Accession # P08709-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.