Learn More
American Research Products Inc Human coagulation factor VIII
Catalog No. 5000328316
Encompass
Human coagulation factor VIII (FVIII) ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 125 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28316 Supplier American Research Products Inc Supplier No. CSBE13861H
Human coagulation factor VIII (FVIII) ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 125 ng/ml Sample serum plasma tissue homogenates Method Sandwich