Learn More
Biomatik Corporation Human Coagulation Factor X (F10) CLIA Kit
Supplier:  Biomatik Corporation EKU09893
Encompass_Preferred
Human Coagulation Factor X (F10) CLIA Kit for detecting F10 in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 8.23-6,000pg/ml. Sensitivity: 3.05pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P00742. Gene ID: 2159. Target Synonyms: FX; FXA; Stuart Prower Factor; Thrombokinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4499