Learn More
Biomatik Corporation Human Coiled Coil And C2 Domain Containing Protein 1A (CC2D1A) ELISA Kit
Supplier:  Biomatik Corporation EKU03291
Encompass_Preferred
Human Coiled Coil And C2 Domain Containing Protein 1A (CC2D1A) ELISA Kit for detecting CC2D1A in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P1N0. Gene ID: 54862. Target Synonyms: FREUD-1; MRT3; AKI1; Mental Retardation,Nonsyndromic,Autosomal Recessive 3; Five prime repressor element under dual repression-binding 1; Akt kinase-interacting protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4369