Learn More
BIOHIPPO Human COL11a2 Microsample
SDP
Catalog No. 5000314878
Encompass
Human COL11a2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of COL11a2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human COL11a2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human COL11a2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human COL11a2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14878 Supplier BIOHIPPO Supplier No. BHE15211365
Human COL11a2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of COL11a2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human COL11a2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human COL11a2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human COL11a2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color