Learn More
BIOHIPPO Human COL4a5 Microsample
SDP
Catalog No. 5000314564
Encompass
Human COL4a5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of COL4a5 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 09 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human COL4a5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human COL4a5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human COL4a5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14564 Supplier BIOHIPPO Supplier No. BHE15211251
Human COL4a5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of COL4a5 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 09 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human COL4a5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human COL4a5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human COL4a5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color