Learn More
BIOHIPPO Human COL6 Microsample
SDP
Catalog No. 5000314556
Encompass
Human COL6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of COL6 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 7 82-500 ng/mL with a sensitivity of 2 8 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human COL6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human COL6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human COL6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14556 Supplier BIOHIPPO Supplier No. BHE15211855
Human COL6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of COL6 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 7 82-500 ng/mL with a sensitivity of 2 8 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human COL6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human COL6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human COL6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color