Learn More
Biomatik Corporation Human Cold Shock Domain Containing Protein E1, RNA Binding (CSDE1) ELISA Kit
Supplier:  Biomatik Corporation EKU03295
Encompass_Preferred
Human Cold Shock Domain Containing Protein E1, RNA Binding (CSDE1) ELISA Kit for detecting CSDE1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75534. Gene ID: 7812. Target Synonyms: UNR; NRU; Upstream of NRAS; N-ras upstream gene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7591