Learn More
American Research Products Inc Human Collagen alpha 1 X cha
Catalog No. 5000327892
Encompass
Human Collagen alpha-1 (X) chain (COL10A1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27892 Supplier American Research Products Inc Supplier No. CSBEL005715HU
Human Collagen alpha-1 (X) chain (COL10A1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich