Learn More
Rockland Immunochemicals Human COLLAGEN Type I - 009-001-103
SDP
Supplier:  Rockland Immunochemicals 009001103
Encompass
Collagen type I is the fibrillar collagen found in most connective tissues, including cartilage. Collagen type I is a protein that strengthens and supports many tissues in the body, including cartilage, bone, tendon, skin and the white part of the eye (sclera).
Catalog No. NC1535766