Learn More
HUMABIOLOGICS INC HumaDerm - Human Skin Collagen Type I, 6mg/ml-10 ml. HumaDerm solution is ideal for 2D & 3D cell culture, organoids, hydrogel, bioinks, and many other regenerative medicine applications. >95% purity.
Supplier:  HUMABIOLOGICS INC SCS610ML
Encompass_Preferred
HumaDerm is the market only native human skin collagen solution for hydrogel, organoids, tissue modeling, bioink, and other regenerative medicine applications. The optimal concentration may vary depending on cell type and application and must be optimized for best results. It is reconstituted in HCl at pH ~2.0 and is supplied at a 6 mg/ml concentration. It is isolated from human skin sourced strictly from American Association of Tissue Banks (AATB) accredited and FDA registered tissue banks and organ procurement organizations (OPOs). HumaDerm is manufactured under ISO 13485 certified and GMP-compliant quality system. HumaDerm is obtained from human tissue that has been tested negative for a minimum of HIV-1 and -2, hepatitis B, and hepatitis C, as well as other infectious agents. HumaDerm is for research use only and is not intended for human use, diagnosis, screening, household, food, or other uses. HumaDerm solution should be stored at 2-8 °C upon receipt. Do not freeze.
Catalog No. NC2107418