Learn More
Rockland Immunochemicals Human COLLAGEN Type VI - 009-001-108
SDP
Supplier:  Rockland Immunochemicals 009001108
Encompass
Collagen type VI is a form of collagen primarily associated with the extracellular matrix of skeletal muscle. Collagen type VI is associated with the genes COL6A1, COL6A2, and COL6A3. Defects in collagen VI are associated with Bethlem myopathy and Ullrich congenital muscular dystrophy.
Catalog No. 50-105-7314