Learn More
Biomatik Corporation Human Collagen Type XIII (COL13) ELISA Kit
Supplier:  Biomatik Corporation EKU03348
Encompass_Preferred
Human Collagen Type XIII (COL13) ELISA Kit for detecting COL13 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5TAT6. Gene ID: 1305. Target Synonyms: COL13A1; COL13-A1; Collagen Alpha-1 (XIII) chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4434