Learn More
Biomatik Corporation Human Colony Stimulating Factor 3, Granulocyte (GCSF) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12321
Encompass_Preferred
Human Colony Stimulating Factor 3, Granulocyte (GCSF) Mini sample ELISA Kit for detecting GCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09919. Gene ID: 1440. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7983