Learn More
Biomatik Corporation Human Colony Stimulating Factor 1, Macrophage (MCSF) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11609
Encompass_Preferred
Human Colony Stimulating Factor 1, Macrophage (MCSF) Mini sample ELISA Kit for detecting MCSF in serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09603. Gene ID: 1435. Target Synonyms: CSF1; M-CSF; Lanimostim; Processed macrophage colony-stimulating factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8120