Learn More
Biomatik Corporation Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF) CLIA Kit
Supplier:  Biomatik Corporation EKU09736
Encompass_Preferred
Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF) CLIA Kit for detecting GM-CSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04141. Gene ID: 1437. Target Synonyms: CSF2; GMCSF; Sargramostim; Molgramostin; Granulocyte-Macrophage Colony Stimulating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5113