Learn More
Sino Biological Human COMMD8 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14666T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human COMMD8 (rh COMMD8; Catalog#14666-H07E; AAH19826.1; Met1-Lys183). COMMD8 specific IgG was purified by Human COMMD8 affinity chromatography.
Catalog No. 50-237-3463