Learn More
Biomatik Corporation Human COMP (Cartilage oligomeric matrix protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58458
Encompass_Preferred
Human COMP (Cartilage oligomeric matrix protein) ELISA Kit for detecting COMP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49747. Target Synonyms: COMP; EDM1; EPD1; MED; PSACH; THBS5; TSP-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5884