Learn More
American Research Products Inc Human Complement C1s subcompon
Catalog No. 5000327129
Encompass
Human Complement C1s subcomponent (C1S) ELISA kit from CUSABIO Detection Range 25 ng/mL-1600 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27129 Supplier American Research Products Inc Supplier No. CSBEL003657HU
Human Complement C1s subcomponent (C1S) ELISA kit from CUSABIO Detection Range 25 ng/mL-1600 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Sandwich