Learn More
Biomatik Corporation Human Complement C2 (C2) Elisa Kit
Supplier:  Biomatik Corporation EKC31198
Encompass_Preferred
Human Complement C2 (C2) Elisa Kit for detecting Complement Component 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-250ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P06681; C2. Target Synonyms: ARMD14; C2; C3/C5 convertase; CO2; CO2_HUMAN; Complement C2; Complement C2a fragment; complement component 2; DKFZp779M0311. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7377