Learn More
Biomatik Corporation Human Complement C3 Convertase (C3 Convertase) CLIA Kit
Supplier:  Biomatik Corporation EKU08912
Encompass_Preferred
Human Complement C3 Convertase (C3 Convertase) CLIA Kit for detecting C3 Convertase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01024. Gene ID: 718. Target Synonyms: C4b2a; C3bBb, complement C.hivin.4.hivin2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8595