Learn More
Abcam Human Complement C3 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1088221X96TESTS
Encompass_Preferred
Human Complement C3 ELISA Kit ab108822 is used to quantify Human Complement C3 with a sensitivity of 8 pg/ml. Competitive ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2045692