Learn More
Abcam Human Complement C3a des Arg ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1330371X96TESTS
Encompass_Preferred
Human Complement C3a des Arg ELISA Kit is a Competitive ELISA kit for the measurement of Human Complement C3a des Arg in Human in Plasma samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2574875