Learn More
ACROBiosystems Human Complement C5 100ug
Catalog No. 502535221
Encompass_Preferred
Human Complement C5 Protein Fc Tag (CO5-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # P01031-1).
Catalog No. 50-253-5221 Supplier ACROBiosystems Supplier No. CO5H5253100UG
Human Complement C5 Protein Fc Tag (CO5-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # P01031-1).