Learn More
Abcam Human Complement C5a ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1936951X96TESTS
Encompass_Preferred
Human Complement C5a ELISA Kit ab193695 is used to quantify Human Complement C5a with a sensitivity of 0.17 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2680902