Learn More
Biomatik Corporation Human Complement Component 1, Q Subcomponent C (C1qC) CLIA Kit
Supplier:  Biomatik Corporation EKU09859
Encompass_Preferred
Human Complement Component 1, Q Subcomponent C (C1qC) CLIA Kit for detecting C1qC in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.03pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02747. Gene ID: 714. Target Synonyms: C1QG; Complement Component 1 q subcomponent,Gamma Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5420