Learn More
Biomatik Corporation Human Complement Component 1, S Subcomponent (C1s) ELISA Kit
Supplier:  Biomatik Corporation EKU03379
Encompass_Preferred
Human Complement Component 1, S Subcomponent (C1s) ELISA Kit for detecting C1s in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09871. Gene ID: 716. Target Synonyms: C1 esterase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9039