Learn More
Biomatik Corporation Human Complement Component 1, Q Subcomponent A (C1qA) ELISA Kit
Supplier:  Biomatik Corporation EKU03374
Encompass_Preferred
Human Complement Component 1, Q Subcomponent A (C1qA) ELISA Kit for detecting C1qA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 35pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02745. Gene ID: 712. Target Synonyms: Complement Component 1,q Subcomponent,Alpha Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9041