Learn More
Biomatik Corporation Human Complement Component 2 (C2) ELISA Kit
Supplier:  Biomatik Corporation EKU03380
Encompass_Preferred
Human Complement Component 2 (C2) ELISA Kit for detecting C2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 2.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06681. Gene ID: 717. Target Synonyms: CO2; C3/C5 convertase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7481