Learn More
Biomatik Corporation Human Complement Component 4c (C4c) CLIA Kit
Supplier:  Biomatik Corporation EKU10046
Encompass_Preferred
Human Complement Component 4c (C4c) CLIA Kit for detecting C4c in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.08ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P0C0L5. Gene ID: 100293534. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4715