Learn More
Biomatik Corporation Human Complement Component 8g (C8g) CLIA Kit
Supplier:  Biomatik Corporation EKU09852
Encompass_Preferred
Human Complement Component 8g (C8g) CLIA Kit for detecting C8g in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.69pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P07360. Gene ID: 733. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6726