Learn More
Biomatik Corporation Human Complement Factor B (CFB) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10534
Encompass_Preferred
Human Complement Factor B (CFB) Wide-range ELISA Kit for detecting CFB in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.44ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00751. Gene ID: 629. Target Synonyms: CF-B; BF; BFD; CFAB; GBG; H2-Bf; PBF2; B-Factor; Properdin; Properdin factor B; C3/C5 convertase; Glycine-rich beta glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6318