Learn More
Biomatik Corporation Human Complement Factor H (CFH) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10536
Encompass_Preferred
Human Complement Factor H (CFH) Wide-range ELISA Kit for detecting CFH in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1,500ng/ml. Sensitivity: 8.52ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08603. Gene ID: 3075. Target Synonyms: CF-H; FH; FHL1; ARMD4; ARMS1; CFHL3; HF1; HF2; HUS; H Factor 2; Age-Related Maculopathy Susceptibility 1; Adrenomedullin binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5278