Learn More
Sino Biological Recombinant Human Factor H Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10714H08H1
Encompass_Preferred
A DNA sequence encoding the Human C-terminal segment of CFH isoform a (NP_000177.2), corresponding to amino acid (Ser 860-Arg 1231) was expressed, fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-224-6817