Learn More
American Research Products Inc Human Complement factor H rela
Catalog No. 5000327821
Encompass
Human Complement factor H-related protein 3 (CFHR3) ELISA kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 8 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27821 Supplier American Research Products Inc Supplier No. CSBEL005276HU
Human Complement factor H-related protein 3 (CFHR3) ELISA kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 8 ng/mL Sample serum plasma tissue homogenates Method Sandwich