Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Complement Factor I (aa 372-516) Control Fragment Recombinant Protein

Catalog No. RP95398
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP95398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP95398 Supplier Invitrogen™ Supplier No. RP95398
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-81923. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05156
Concentration 1.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 3426
Name Human Complement Factor I (aa 372-516) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I
Common Name Complement Factor I
Gene Symbol CFI
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GCWILTAAHCLRASKTHRYQIWTTVVDWIHPDLKRIVIEYVDRIIFHENYNAGTYQNDIALIEMKKDGNKKDCELPRSIPACVPWSPYLFQPNDTCIVSGWGREKDNERVFSLQWGEVKLISNCSKFYGNRFYEKEMECAGTYDG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.