Learn More
Biomatik Corporation Human Complement Factor P (CFP) CLIA Kit
Supplier:  Biomatik Corporation EKU09846
Encompass_Preferred
Human Complement Factor P (CFP) CLIA Kit for detecting CFP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P27918. Gene ID: 5199. Target Synonyms: CF-P; PFC; Properdin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7776