Learn More
American Research Products Inc Human Complement fragment 3b
Catalog No. 5000326772
Encompass
Human Complement fragment 3b C3b ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum urine cerebrospinal fluid (CSF) saliva Method Sandwich
Catalog No. 50-003-26772 Supplier American Research Products Inc Supplier No. CSBE09303H
Human Complement fragment 3b C3b ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum urine cerebrospinal fluid (CSF) saliva Method Sandwich