Learn More
American Research Products Inc Human Complement fragment 5a
Catalog No. 5000329491
Encompass
Human Complement fragment 5a C5a ELISA Kit from CUSABIO Detection Range 39 pg/ml-2500 pg/ml Sensitivity 9 75 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29491 Supplier American Research Products Inc Supplier No. CSBE08512H
Human Complement fragment 5a C5a ELISA Kit from CUSABIO Detection Range 39 pg/ml-2500 pg/ml Sensitivity 9 75 pg/ml Sample serum plasma tissue homogenates Method Sandwich