Learn More
Biomatik Corporation Human Complement Receptor 2 (CD21) ELISA Kit
Supplier:  Biomatik Corporation EKU03432
Encompass_Preferred
Human Complement Receptor 2 (CD21) ELISA Kit for detecting CD21 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20023. Gene ID: 1380. Target Synonyms: CR2; C3DR; Complement Component (3d/Epstein Barr Virus) Receptor 2; Complement C3d receptor; Epstein-Barr virus receptor; EBV receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6208