Learn More
Quidel Corporation Human Serum Standard Complement
SDP
Supplier:  Quidel Corporation A100
Encompass
Uniform pool of human serum complement that has been characterized for levels and activities of a variety of complement proteins.
Catalog No. NC0506946